Trimethyloxonium tetrafluoroborate ≥95%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:420-37-1
:
Formula:C₃H₉BF₄O
MDL Number:MFCD00011798
:147.91 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:206-994-4
UN:3261
:GHS05
Storage Temperature:Freezer