3-Chloropropyldimethylammonium chloride
Catalog # MOLE61642496-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:158.07 g/mol
EINECS:226-467-2
CAS Number:5407-04-5
Formula:C₅H₁₃Cl₂N
MDL Number:MFCD00012521