Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Brij® 35
Brij® 35
Brij® 35
Brij® 35
Supplier:
Molekula
Certificates
Expand image
Brij® 35
Supplier:
Molekula
About this item
CAS Number:
9002-92-0
:
Formula:
C₅₈H₁₁₈O₂₄
MDL Number:
MFCD00080891
:
1199.56 g/mol
EINECS:
500-002-6
:
GHS07
Storage Temperature:
Ambient