
Trimethyloxonium tetrafluoroborate ≥95.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Artikelnr. 8.41638.0010
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Assay (acidimetric):≥ 95.0 %
- Identity (IR):passes test
- Molar Mass:147.9 g/mol
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Specification Test Results
About this item
:
:147.91 g/mol
ADR:4.3 (8),I
:M-Clarity™ quality level = MQ200
EINECS:206-994-4
UN:3131
:GHS05
Storage Temperature:Freezer
CAS Number:420-37-1
Formula:C₃H₉BF₄O
MDL Number:MFCD00011798