DL-Tropic acid, Sigma-Aldrich®
8.08384.0025
: Merck
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Solubility:20 g/l (20 °C)
- Melting Point:115 - 118 °C
- Molar Mass:166.17 g/mol
:
:166.18 g/mol
:M-Clarity™ quality level = MQ200
:208-465-3
:529-64-6
:C₉H₁₀O₃
:MFCD00004255