1-(4-Methoxybenzyl)piperazine
Catalog # FLUO021565-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:206.29 g/mol
CAS Number:21867-69-6
Formula:C₁₂H₁₈N₂O
MDL Number:MFCD01075236