Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclohexane

Cyclohexane

Glass bottle, 1 L
Catalog # KRAE27-050.1000
Flammable
Health
Irritant
Environment
Supplier:   KRAEMER MARTIN
Cyclohexane
Catalog # KRAE27-050.1000
Supplier:  KRAEMER MARTIN
CAS Number:  110-82-7
Cyclohexane
Cyclohexane

Glass bottle, 1 L

CAS Number:  110-82-7
Price
30,60 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.