Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1'-Bis(diphenylphosphino)ferrocene
1,1'-Bis(diphenylphosphino)ferrocene
1,1'-Bis(diphenylphosphino)ferrocene
1,1'-Bis(diphenylphosphino)ferrocene
Supplier:
COMBI-BLOCKS
Certificates
Expand image
1,1'-Bis(diphenylphosphino)ferrocene
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
12150-46-8
:
Formula:
C₃₄H₂₈FeP₂
MDL Number:
MFCD00001422
:
554.39 g/mol
EINECS:
430-420-3