Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Phenyl-3-formylpyridine
6-Phenyl-3-formylpyridine
6-Phenyl-3-formylpyridine
6-Phenyl-3-formylpyridine
Supplier:
BIONET RESEARCH
Certificates
Expand image
6-Phenyl-3-formylpyridine
Supplier:
BIONET RESEARCH
About this item
CAS Number:
63056-20-2
:
Formula:
C₁₂H₉NO
MDL Number:
MFCD03086130
:
183.21 g/mol