TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Catalog # A1084.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:229.25 g/mol
EINECS:230-906-3
:GHS07
Storage Temperature:Ambient
CAS Number:7365-44-8
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532