1,1'-Bis(diphenylphosphino)ferrocene ≥97%
Catalog # B21166.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:554.39 g/mol
Melting Pt:∼ 182 °C (dec.)
EINECS:430-420-3
:GHS07
CAS Number:12150-46-8
Formula:C₃₄H₂₈FeP₂
MDL Number:MFCD00001422