Order Entry
Germany
Orders LinkContactUsLinkComponent
Brij® 35

Brij® 35

Brij® 35
Certificates
Brij® 35

3 Options Available Below

Product Details & Documents

CAS No.: 9002-92-0 Molecular Formula: C58H118O24 Formula Weight: 1199.57 Physical Form: Waxy solid Appearance: White Melting Point: ca 35-40° MDL No.: MFCD00080891

For use in Stein-Moore chromatography

Synonyms:  Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
  • CAS Number:9002-92-0
  • Formula:C₅₈H₁₁₈O₂₄
  • MDL Number:MFCD00080891
  • Molecular Weight:1199.56 g/mol
  • Melting Pt:∼ 35…40 °C
  • Storage Temperature:Ambient
  • EINECS:500-002-6
Product Family Options

Product Information