Order Entry
Germany
ContactUsLinkComponent
 
 
Trimethyloxonium tetrafluoroborate 95%
 
undefined
 
Trimethyloxonium tetrafluoroborate 95%

 

  • Appearance:
    White to off-white Powder or crystals and/or chunks
  • Titration with NaOH:
    94.0 to 106.0 %
  • Refractive index:
    1.4420 to 1.4450 (20°C, 589 nm)
  • GC:
    ≥88 %