Order Entry
Germany
ContactUsLinkComponent
 
 
3-Formyl-6-methoxypyridine 97%
 
undefined
 
3-Formyl-6-methoxypyridine 97%

 

  • Appearance:
    Off-white to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    46 to 54 °C
  • GC:
    ≥96.0 %