Saccharin sodium hydrate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82385-42-0
:MFCD00151213
Formula:C₇H₅NNaO₃S·xH₂O
MDL Number:MFCD00149605
:205.17 g/mol (anhy)
Storage Temperature:Ambient
EINECS:204-886-1
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:≤15 %
- Additional info:Benzoate and salicylate: passes test
- GC:≤25 ppm Toluenesulfonamides
- Readily carbonizable substance:Passes test
- Titration with HClO4:99.0 to 101.0 % (on anhydrous substance)
- Arsenic (As):≤3 ppm
- Selenium (Se):≤30 ppm
- Heavy metals (as Pb):≤20 ppm