TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) 99% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7365-44-8
:
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532
:229.25 g/mol
Melting Pt:223…225 °C
EINECS:230-906-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Solubility:(0.1 M in water) Clear colorless
- UV:(0.1 M in water)
- pH:4 to 8 (0.1 M at 20°C)
- Titration with NaOH:≥98.5 %
- Heavy metals (as Pb):≤5 ppm