
2-Chloropropyldimethylammonium chloride 98.5%
ACRO115835000
:
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Titration with HClO4:≥98.0 % (On dry substance)
:4584-49-0
:
:C₅H₁₃Cl₂N
:MFCD00012534
:158.07 g/mol
:191…195 °C
:224-971-7
:GHS07
:Ambient