α-Acetyl-γ-butyrolactone 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:517-23-7
:
Formula:C₆H₈O₃
MDL Number:MFCD00005394
:128.13 g/mol
Boiling Pt:107…108 °C (5 mmHg)
Density:1.18 g/cm³
Flash Pt:140 °C
EINECS:208-235-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4580 to 1.4610 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤30 APHA