Cyclohexane ≥99% for HPLC
About this item
Purity: 99% min CAS No.: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -18°(-1°F) Physical Form: Liquid Density: 0.779 Refractive Index: 1.4260 Appearance: Colourless Odour: Pungent Melting Point: 6.5° Boiling Point: 80-81° Beilstein Reference: 1900225 Merck Reference: 14,2723 UN No.: 1145 MDL No.: MFCD00003814
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:14,2723
- Boiling Pt:80…81 °C
- Melting Pt:6.5 °C
- Density:0.779
- Flash Pt:–18 °C (–1 °F)
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
2 Options Available Below
Product Details & Documents
Purity: 99% min CAS No.: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -18°(-1°F) Physical Form: Liquid Density: 0.779 Refractive Index: 1.4260 Appearance: Colourless Odour: Pungent Melting Point: 6.5° Boiling Point: 80-81° Beilstein Reference: 1900225 Merck Reference: 14,2723 UN No.: 1145 MDL No.: MFCD00003814
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:14,2723
- Boiling Pt:80…81 °C
- Melting Pt:6.5 °C
- Density:0.779
- Flash Pt:–18 °C (–1 °F)
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



