
Cyclohexanecarboxylic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-89-5
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00001461
:128.17 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:30…32 °C
Density:1.03 g/cm³
Flash Pt:>112 °C
EINECS:202-711-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Fused crystalline solid or viscous liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
- Elemental Analysis:33.33 to 35.03 % C