Order Entry
Germany
ContactUsLinkComponent
Cyclopentanone for synthesis, Sigma-Aldrich®
Flammable
Irritant
Cyclopentanone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Cyclopentanone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    -58 °C
  • Molar Mass:
    84.11 g/mol
  • Boiling Point:
    131 °C (1013 hPa)
  • Vapor Pressure:
    11.5 hPa (20 °C)
  • Flash Point:
    27 °C
  • Explosion Limit:
    1.6 - 10.8 %(V)
  • Density:
    0.95 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    445 °C
  • Hill Formula:
    C5H8O

Specification Test Results

Related Information

Certificates
Search by Lot Number