

Cyclopentanone 99+%, pure
:120-92-3
:
:C₅H₈O
:MFCD00001409
:84.12 g/mol
:13,02771
:130…131 °C (760 mmHg)
:–51 °C
:0.95 g/cm³
:26 °C
:Ambient
:204-435-9
:2245
:3,III
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4380 (20°C, 589 nm)
- GC:≥99.0 %
- Stabilizer:0.01 % alpha-Tocopherol