Cyclooctanone
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:502-49-8
- Formula:C₈H₁₄O
- MDL Number:MFCD00001754
- Molecular Weight:126.2 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:39…41 °C
- Density:0,9581 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-940-2
- UN:1759
- ADR:8,III
3 Options Available Below
Product Details & Documents
- CAS Number:502-49-8
- Formula:C₈H₁₄O
- MDL Number:MFCD00001754
- Molecular Weight:126.2 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:39…41 °C
- Density:0,9581 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-940-2
- UN:1759
- ADR:8,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



