
Bromocyclohexane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-85-0
:
Formula:C₆H₁₁Br
MDL Number:MFCD00003819
:163.06 g/mol
Merck Index:14,2730
Boiling Pt:166…167 °C
Melting Pt:–57 °C
Density:1.335
Flash Pt:62 °C (143 °F)
EINECS:203-622-2
:GHS07
Storage Temperature:Ambient