Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methylquinaldine
6-Methylquinaldine
6-Methylquinaldine
6-Methylquinaldine
FLUO018867-10G
:
FLUOROCHEM
:
6-Methylquinaldine
FLUO018867-10G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
157.22 g/mol
:
266 °C (1013 hPa)
:
55…58 °C
:
Ambient
:
212-891-5
:
877-43-0
:
C₁₁H₁₁N
:
MFCD00006762