Quinoline ≥97%
About this item
Purity: 97% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Refractive Index: 1.6260 Melting Point: -15° Boiling Point: 237° Solubility: More soluble in hot than cold water. Miscible with alcohol, ether Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:14,8068
- Boiling Pt:237 °C
- Melting Pt:–15 °C
- Density:1.090
- Flash Pt:101 °C (214 °F)
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
4 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Refractive Index: 1.6260 Melting Point: -15° Boiling Point: 237° Solubility: More soluble in hot than cold water. Miscible with alcohol, ether Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:14,8068
- Boiling Pt:237 °C
- Melting Pt:–15 °C
- Density:1.090
- Flash Pt:101 °C (214 °F)
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



