Order Entry
Germany
Orders LinkContactUsLinkComponent
Quinoline 96%

Quinoline 96%

Toxic
Health
Environment
Quinoline 96%
Certificates
Quinoline 96%

4 Options Available Below

Product Details & Documents
  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:13,08160
  • Boiling Pt:237 °C (760 mmHg)
  • Melting Pt:–15 °C
  • Density:1.095 g/cm³
  • Flash Pt:101 °C
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear light yellow to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6230 to 1.6270 (20°C, 589 nm)
  • GC:
    ≥95 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 74,80 €
    74,80 €
    1 * 500 mL
       74,80 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 119,00 €
    119,00 €
    1 * 1 L
       119,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 204,00 €
    204,00 €
    1 * 2,5 L
       204,00 €

  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 638,00 €
    638,00 €
    1 * 10 L
       638,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.