Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Bromoquinoline

3-Bromoquinoline

25 g
Catalog # MOLE12350675-25G
Irritant
Marketsource
Supplier:   Molekula
3-Bromoquinoline
Catalog # MOLE12350675-25G
Supplier:  Molekula
Marketsource
CAS Number:  5332-24-1
3-Bromoquinoline

25 g

CAS Number:  5332-24-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:5332-24-1
  • Formula:C₉H₆BrN
  • MDL Number:MFCD00006767
  • Molecular Weight:208.06 g/mol
  • Boiling Pt:274 °C (1013 hPa)
  • Melting Pt:15 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:226-237-1
Specifications

Specifications

  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.