1-Phenylcyclohexanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1589-60-2
:
Formula:C₁₂H₁₆O
MDL Number:MFCD00021393
:176.26 g/mol
Boiling Pt:153 °C (20 mmHg)
Melting Pt:58…62 °C
Density:1.101 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-456-0