Orders LinkContactUsLinkComponent
Saccharometers
Saccharometers
Supplier:   GERING
Saccharometers
Supplier:  GERING
Certificates
Saccharometers
6 Options Available Below
Product Details & Documents
These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.
Product Family Options
Product Information
  • Range
    Division
    Temperature Range
    Availability
    Price
  • 0 - 7 %
    0,1
    0…30 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,00 €
    97,00 €
    1 * 1 ST
       97,00 €
  • 0 - 20 %
    0,1
    4…26 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
  • 0 - 24 %
    0,1
    4…26 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 287,00 €
    287,00 €
    1 * 1 ST
       287,00 €
  • 10 - 20%
    0,1
    4…26 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 256,00 €
    256,00 €
    1 * 1 ST
       256,00 €
  • 10 - 25 %
    0,1
    4…26 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 256,00 €
    256,00 €
    1 * 1 ST
       256,00 €
  • 15 - 20 %
    0,1
    5…25 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €