Order Entry
Germany
Orders LinkContactUsLinkComponent
2,4,6-Trimethylaniline

2,4,6-Trimethylaniline

Irritant
Marketsource
Supplier:   CHEMPUR
2,4,6-Trimethylaniline
Supplier:  CHEMPUR
Marketsource
Certificates
2,4,6-Trimethylaniline

2 Options Available Below

Product Details & Documents
  • CAS Number:88-05-1
  • Formula:C₉H₁₃N
  • MDL Number:MFCD00007740
  • Molecular Weight:135.21 g/mol
  • Boiling Pt:233 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:0,96 g/cm³ (20 °C)
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
  • EINECS:201-794-3
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.