Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2,4,6-Trimethylaniline ≥98%

Toxic
Irritant
2,4,6-Trimethylaniline ≥98%
Certificates
2,4,6-Trimethylaniline ≥98%
2,4,6-Trimethylaniline ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 88-05-1 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 96°(204°F) Refractive Index: 1.5510 Melting Point: -5° Boiling Point: 232-234° Beilstein Reference: 636559 UN No.: 2810 MDL No.: MFCD00007740

  • CAS Number:88-05-1
  • Formula:C₉H₁₃N
  • MDL Number:MFCD00007740
  • Molecular Weight:135.21 g/mol
  • Boiling Pt:232…234 °C
  • Melting Pt:–5 °C
  • Density:0.962
  • Flash Pt:96 °C (204 °F)
  • Storage Temperature:Ambient
  • EINECS:201-794-3
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information