4-tert-Butyltoluene 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-51-1
:
Formula:C₁₁H₁₆
MDL Number:MFCD00008837
Boiling Pt:189…192 °C (760 mmHg)
Melting Pt:–54 °C
Density:0.8612 g/cm³
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:202-675-9
ADR:6.1,II
:148.25 g/mol
UN:2929
:GHS06
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4910 to 1.4930 (20°C, 589 nm)
- GC:≥95 %