Waste bin, B1, Tork®
Catalog # 115-2423
Supplier: Essity
Specifications
- Capacity:50 L
- Colour:White
- Seq:01
- W×D×H:341×258×706 mm
- Pk:1
Specifications
About this item
This white metal bin is suitable for non hazardous waste.
- Free standing or wall-mounted: Flexible
- Easy to fit the lid: Holds the bin liner in place