ContactUsLinkComponent
Cytology funnels, CytoSep™ for StatSpin Cytofuge® 2 cytocentrifuge
Cytology funnels, CytoSep™ for StatSpin Cytofuge® 2 cytocentrifuge
Catalog # 720-1972
Supplier:  Simport Scientific
undefined
Cytology funnels, CytoSep™ for StatSpin Cytofuge® 2 cytocentrifuge
Catalog # 720-1972
Supplier:  Simport Scientific
Specifications
  • For:
    StatSpin Cytofuge® 2 cytocentrifuge
  • Pk:
    48
Specifications
About this item
CytoSep™ cytology funnels for the StatSpin Cytofuge® 2 cytocentrifuge allow cells to gently concentrate in a 7 mm diameter area on the slide, while supernatant is simultaneously absorbed by a filter card. Will accept 50 to 400 μl of sample. Produces cell mono layer presentations of excellent quality.