ContactUsLinkComponent
Flexible couplings for stirrers
Flexible couplings for stirrers
Catalog # LENZ06400000
undefined
Flexible couplings for stirrers
Catalog # LENZ06400000
Specifications
  • Description:
    Flexible coupling for stirrers, with pivot 8 - 10 mm
  • Pk:
    1
Specifications
About this item
Flexible coupling made of stainless steel.
  • Compensates for axial and height differences
  • No adjustments necessary for motor and stirring rod
  • Stainless steel clamping chuck with clamping range 8 to 10 mm