

2,6-Di-tert-butylphenol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:128-39-2
:
Formula:C₁₄H₂₂O
MDL Number:MFCD00008820
Boiling Pt:253 °C (760 mmHg)
Melting Pt:35…38 °C
Density:0,914 g/cm³ (20 °C)
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:204-884-0
ADR:9,III
:206.33 g/mol
UN:3077
:GHS09
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:35 to 38 °C
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %