2-Iodo-p-xylene ≥98%
Supplier: Thermo Fisher Scientific
Wechseln zu:
Über diesen Artikel
Purity: 98+% CAS No.: 1122-42-5 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 229-230° Beilstein Reference: 1905728 MDL No.: MFCD00013708
Synonyme: 2-Iodo-1,4-dimethylbenzene, 1,4-Dimethyl-2-iodobenzene
- CAS-Nummer:1122-42-5
- Formel::C₈H₉I
- MDL-Nummer:MFCD00013708
- Molekulargewicht:232.06 g/mol
- Siedepunkt:229…230 °C
- Dichte:1.617
- Flammpunkt:>110 °C (230 °F)
- Lagertemperatur:Ambient
- EINECS:214-347-2
2 mögliche Produktvariationen vorhanden
Product Details & Documents
Purity: 98+% CAS No.: 1122-42-5 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 229-230° Beilstein Reference: 1905728 MDL No.: MFCD00013708
Synonyme: 2-Iodo-1,4-dimethylbenzene, 1,4-Dimethyl-2-iodobenzene
- CAS-Nummer:1122-42-5
- Formel::C₈H₉I
- MDL-Nummer:MFCD00013708
- Molekulargewicht:232.06 g/mol
- Siedepunkt:229…230 °C
- Dichte:1.617
- Flammpunkt:>110 °C (230 °F)
- Lagertemperatur:Ambient
- EINECS:214-347-2
Documents
Organic Building Blocks and Intermediates



