Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chlorodiisopropylsilane
Chlorodiisopropylsilane
Chlorodiisopropylsilane
Chlorodiisopropylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Chlorodiisopropylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
2227-29-4
:
Formula:
C₆H₁₅ClSi
MDL Number:
MFCD00054896
:
150.72 g/mol