Urea 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIALU7129-1VL
Supplier: Merck
CAS Number:
Specifications
- Pk:1 Vial
- Seq:0032
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022