Perfluorodecahydronaphthalene (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIALP9900-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:462.08
:M-Clarity™ quality level = MQ100
EINECS:206-192-4
CAS Number:306-94-5
Formula:C₁₀F₁₈
MDL Number:MFCD00010626