Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lidocaine hydrochloride monohydrate
Lidocaine hydrochloride monohydrate, Sigma-Aldrich®
Lidocaine hydrochloride monohydrate, Sigma-Aldrich®
Lidocaine hydrochloride monohydrate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Lidocaine hydrochloride monohydrate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
6108-05-0
:
Formula:
C₁₄H₂₃ClN₂O·1H₂O
MDL Number:
MFCD00150329
:
270.8 g/mol
ADR:
6.1,III
:
M-Clarity™ quality level = MQ200
UN:
2811