Order Entry
Germany
ContactUsLinkComponent
Tetrabutylammonium hydrogen sulphate 55% in aqueous solution, Sigma-Aldrich®
Tetrabutylammonium hydrogen sulphate 55% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL72453-250ML-F
Supplier:  Merck
CAS Number:  
Tetrabutylammonium hydrogen sulphate 55% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL72453-250ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :339.54 g/mol
EINECS:251-068-5
CAS Number:32503-27-8
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637