Order Entry
Germany
ContactUsLinkComponent
Benzofurazan, Sigma-Aldrich®
Benzofurazan, Sigma-Aldrich®
Catalog # SIAL650137-1G
Supplier:  Merck
CAS Number:  
Benzofurazan, Sigma-Aldrich®
Catalog # SIAL650137-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :120.11 g/mol
CAS Number:273-09-6
Formula:C₆H₄N₂O
MDL Number:MFCD00068064