Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
23190-16-1
:
Formula:
C₁₄H₁₅NO
MDL Number:
MFCD00074960
:
213.28 g/mol