Order Entry
Germany
Orders LinkContactUsLinkComponent
Galactitol

Galactitol

Glass bottle for solids, 100 g
Catalog # MOLEM42994090
Marketsource
Supplier:   Molekula
Galactitol
Catalog # MOLEM42994090
Supplier:  Molekula
Marketsource
CAS Number:  608-66-2
Galactitol

Glass bottle for solids, 100 g

CAS Number:  608-66-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient
  • EINECS:210-165-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.