Order Entry
Germany
ContactUsLinkComponent
Tetramethylammonium hydrogen sulphate
Tetramethylammonium hydrogen sulphate
Catalog # MOLE27719041
Supplier:  Molekula
CAS Number:  
Tetramethylammonium hydrogen sulphate
Catalog # MOLE27719041
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.22 g/mol
EINECS:279-490-5
CAS Number:80526-82-5
Formula:C₄H₁₃NO₄S
MDL Number:MFCD00036149