Isopentyl nitrite (mixed isomers ) stabilised, Sigma-Aldrich®
: Merck
:110-46-3
:
:C₅H₁₁NO₂
:MFCD00002057
:117.15 g/mol
:3,II
:M-Clarity™ quality level = MQ200
:Stabilised with potassium carbonate
:203-770-8
:1113
:GHS02
:GHS06
:Ambient
- Solubility:(20 °C) practically insoluble
- Molar Mass:117.15 g/mol
- Boiling Point:97 - 99 °C (1013 hPa)
- Flash Point:-18 °C
- Density:0.872 g/cm3 (20 °C)
- Ignition Point:208 °C
- Synonyms:iso-Amyl nitrite, iso-Pentyl nitrite, Nitrous acid isoamyl ester
- Hill Formula:C5H11NO2