Order Entry
Germany
ContactUsLinkComponent
Hydrochloric acid 2 mol/l (2 N) for TOC
Corrosive
Hydrochloric acid 2 mol/l (2 N) for TOC
Catalog # KRAF12494.1000
Supplier:  Bernd Kraft
CAS Number:  
Corrosive
Hydrochloric acid 2 mol/l (2 N) for TOC
Catalog # KRAF12494.1000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324