0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-(2-Bromoethoxy)-4-methoxybenzene
1-(2-Bromoethoxy)-4-methoxybenzene
1-(2-Bromoethoxy)-4-methoxybenzene
1-(2-Bromoethoxy)-4-methoxybenzene
Supplier:
BIONET RESEARCH
Certificates
Expand image
1-(2-Bromoethoxy)-4-methoxybenzene
Supplier:
BIONET RESEARCH
About this item
CAS Number:
22921-76-2
:
Formula:
C₉H₁₁BrO₂
MDL Number:
MFCD00017893
:
231.09 g/mol